Apolipoprotein E/β-amyloid (ApoE/Aβ) complex ELISA kit
Toimittaja: Biosensis
BEK-2224-1P BEK-2224-2P
BSENBEK-2224-1PEA 398 EUR
BSENBEK-2224-1P BSENBEK-2224-2P
Apolipoprotein E/β-amyloid (ApoE/Aβ) complex ELISA kit
Bio-assayt ELISA-kitit
The Biosensis ApoE/Aβ Complex ELISA kit is a sandwich ELISA that allows the preferential measurement of ApoE/Aβ complexes. This kit consists of a pre-coated mouse monoclonal anti-Aβ capture antibody, a highly validated ApoE/Aβ complex standard that is pre-formed, lyophilised and ready for reconstitution, a biotinylated ApoE detection antibody, and horseradish peroxidase (HRP)-conjugated streptavidin and detection reagent. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product which is directly proportional to the level of ApoE/Aβ complex present in samples and protein standards. Importantly, a well-characteriaed and unique ApoE/Aβ complex is included as a standard. This complex is pre-formed and lyophilised, requiring only reconstitution with assay diluent prior to use.
The purpose of this kit is the in vitro qualitative measurement of ApoE/Aβ complexes in brain extracts and CSF samples from both transgenic mice and humans or primates, relative to a known ApoE/Aβ complex standard. This kit has not been tested for other sample applications.
Toimitustiedot: Each kit contains 1 or 2× 96 pre-coated strip plates, protein standards, detection reagents, substrate buffer and precise instructions.