Anti-Parvin alpha Rabbit Polyclonal Antibody

Toimittaja: Boster Bio
PB9555
BSBTPB9555EA 552 EUR
BSBTPB9555
Anti-Parvin alpha Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Alpha-parvin(PARVA) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: Parvin alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR