Anti-HRH3 Rabbit Polyclonal Antibody

Toimittaja: Boster Bio
PA1204
BSBTPA1204EA 552 EUR
BSBTPA1204
Anti-HRH3 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit IgG polyclonal antibody for Histamine H3 receptor (HRH3) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: HRH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR