PicoKine Human IDS/Iduronate 2 Sulfatase; ELISA Kit

Toimittaja: Boster Bio

EK1452
BSBTEK1452EA 646 EUR
BSBTEK1452
PicoKine Human IDS/Iduronate 2 Sulfatase; ELISA Kit
Bio-assayt ELISA-kitit

Sandwich high sensitivity ELISA kit for quantitative detection of human IDS.


  • Sensitivity, detect single digit picogram level of target protein
  • Specificity, thoroughly tested for cross-reactivity with family members
  • Stability, guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, on-demand technical support directly from ELISA experts


Detect Human Iduronate 2-Sulfatase/IDS with <15 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Iduronate 2-Sulfatase/IDS tissue specificity: Liver, kidney, lung, and placenta.


96wells/kit, with removable strips.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR