Volumetric flasks, class A, brown markings, BLAUBRAND® ETERNA
Toimittaja: Brand
DAKKS36849 DAKKS36842 DAKKS36847 DAKKS36952 DAKKS36851 DAKKS36853 DAKKS36845 DAKKS36852
BRNDDAKKS36849EA 131 EUR
BRNDDAKKS36849 BRNDDAKKS36842 BRNDDAKKS36847 BRNDDAKKS36952 BRNDDAKKS36851 BRNDDAKKS36853 BRNDDAKKS36845 BRNDDAKKS36852
Volumetric flasks, class A, brown markings, BLAUBRAND® ETERNA
Kolvit ja erlenmeyerit Mittapullot
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, IN)
  • Mark and inscriptions in ETERNA amber stain
  • DE-M marking
  • Supplied with USP individual certificate or with DAkkS calibration certificate
DIN EN ISO 1042
Sertifikaatit: DIN EN ISO 1042