Reagent Reservoirs with Cover
Toimittaja: Brand
701460 701462 701464
BRND701460EA 140 EUR
BRND701460 BRND701462 BRND701464
Reagent Reservoirs with Cover
Reagenssisäiliöt
For manual pipetting and automation. PP, 40 ml working volume, V-bottom, low dead volume, graduated.
  • CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials
  • Manufactured under controlled room conditions
  • Intensive in-process controls
  • Visual inspections
  • Batch management
  • Final product inspection
  • Proven functionality