Sterilisation indicators
Toimittaja: Brand
61750
BRND61750EA 20.8 EUR
BRND61750
Sterilisation indicators
Sterilointi-indikaattori
This self-adhesive crepe indicator tape turns from white to brown upon autoclaving.