Värjäyskaukalo
Toimittaja: Brand
474300 474305
BRND474300EA 234 EUR
BRND474300 BRND474305
Värjäyskaukalo
Astiat ja purkit Värjäystölkki
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Tilaustiedot: Trough and tray (insert) have to be ordered separately.