Jauhesuppilot, joissa lyhyt leveä varsi
Toimittaja: Brand
146506 146508 146511 146516 146519 146523
BRND146506EA 14.4 EUR
BRND146506 BRND146508 BRND146511 BRND146516 BRND146519 BRND146523
Jauhesuppilot, joissa lyhyt leveä varsi
Suppilot Jauhesuppilot
Borosilicate glass 3,3.
  • Chemically resistant glass
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Quick and easy to clean
Ideal for transferring powders, can also be used for liquids.