Pesupullo, leveä suuaukko, printatulla nimellä
Toimittaja: Brand
144078 144079 144084 144085 144088
BRND144078EA 0 EUR
BRND144078 BRND144079 BRND144084 BRND144085 BRND144088
Pesupullo, leveä suuaukko, printatulla nimellä
Pullot Pesupullot
LDPE, translucent.
  • Large opening for convenient filling
  • Without or with venting valve (for destilled water and organic solvents only) available
  • Colour-coded stoppers
High-quality wash bottles with integrated wash stream and sealing cone closure. Label with designation of contents and general safety statements in English (with CAS numbers and NFPA codes and German (with CAS numbers).