Pipe marker, water
Toimittaja: Brady
N003349 N005832 N005873 N006135 N006160 N006279
BRDYN003349EA 21.4 EUR
BRDYN003349 BRDYN005832 BRDYN005873 BRDYN006135 BRDYN006160 BRDYN006279
Pipe marker, water
Merkit, kyltit ja lattiamerkinnät
Individual markers on card with precut arrows, without pictogram.
  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm