Safety padlocks, long body
Toimittaja: Brady
834462 834464 834465 834463 834466 834467
BRDY834462EA 176 EUR
BRDY834462 BRDY834464 BRDY834465 BRDY834463 BRDY834466 BRDY834467
Safety padlocks, long body
Turvalukot
These nylon padlocks with steel shackle have the added advantage of being fully electrical resistant. The nylon shackle cuts off the electrical current from the shackle to the key.
  • Unique cylinder is insulated to protect workers from shocks when key is inserted
  • Cylinder is 6-pin precision-machined for more unique key cuts and better tamper resistance
  • Reserved, paracentric keyway provides optimal security
  • Special design prohibits the key from being released until the padlock shackle is closed
  • Locks are compact and lightweight and resist impact, temperature,