Thermal transfer printer labels, type B-492, FreezerBondz
Toimittaja: Brady
805920 805913 805915 805919 805918 805912 805911 805917 805916 116373 142076 116388 805914 804786 142078 142077 114288 804788 116389 114287 142075 114289 134098 134097 198751
BRDY805920EA 534 EUR
BRDY805920 BRDY805913 BRDY805915 BRDY805919 BRDY805918 BRDY805912 BRDY805911 BRDY805917 BRDY805916 BRDY116373 BRDY142076 BRDY116388 BRDY805914 BRDY804786 BRDY142078 BRDY142077 BRDY114288 BRDY804788 BRDY116389 BRDY114287 BRDY142075 BRDY114289 BRDY134098 BRDY134097 BRDY198751
Thermal transfer printer labels, type B-492, FreezerBondz
Etiketit, tarrat
FreezerBondz thermal transfer printers labels can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen. These labels are on a 1" core, a 'smaller core' than the majority of labels. Recommended printer for these is the BBP11-34L printer.
  • Good print smudge resistance
  • Solvent resistance
  • Good low temperature performance
  • Widely used in liquid nitrogen and autoclave applications