Label Printer Kits, S3700
Toimittaja: Brady
321264 321265 321266 321267 321268 321269
BRDY321264EA 4990 EUR
BRDY321264 BRDY321265 BRDY321266 BRDY321267 BRDY321268 BRDY321269
Label Printer Kits, S3700
Tulostimet ja tulostimien lisävarusteet Label Printers
Multicolour, cut sign and label printers, S3700 with QWERTZ keyboard, Wifi, bluetooth enabled and BWS SFIDS included. Colour your way to an improved workplace with the S3700 Sign and Label Printer. With multicolour print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
  • Multicolour print capabilities make it ideal for creating impactful safety signs, lockout tags, warning labels and more in a single pass
  • Built in XY-cutter allows for the creation of die-cut shapes and text that