Bulk Linerless Polyester Cable Tags for M611, M610, BMP61, M710, and BMP71
Toimittaja: Brady
312144 312167 312150 312165 312146 312164 312162 312151 312145 312155 312169 312142 312166 312143 312152 312160 312158 312159 312156 312148 312157 312149 312153 312163
BRDY312144EA 553 EUR
BRDY312144 BRDY312167 BRDY312150 BRDY312165 BRDY312146 BRDY312164 BRDY312162 BRDY312151 BRDY312145 BRDY312155 BRDY312169 BRDY312142 BRDY312166 BRDY312143 BRDY312152 BRDY312160 BRDY312158 BRDY312159 BRDY312156 BRDY312148 BRDY312157 BRDY312149 BRDY312153 BRDY312163
Bulk Linerless Polyester Cable Tags for M611, M610, BMP61, M710, and BMP71
Etiketit, tarrat
The B-7598 polyester cable identification tag has been developed to keep cables identified in outdoor environments.
  • Gloss finish
The B-7598 polyester cable identification tags offer up to 10 year outdoor legibility and durability, and are available in six colours. Applications include data and telecommunications labelling, and wire and cable labelling.
Sertifikaatit: Tags supplied in white, black, or silver are halogen-free per IEC 61249-2-21 (2003-11).