Thermoplastic Polyether Polyurethane Tags for M610 and M611
Toimittaja: Brady
312089 312087 312086 312090 312085 312084 312088
BRDY312089EA 107 EUR
BRDY312089 BRDY312087 BRDY312086 BRDY312090 BRDY312085 BRDY312084 BRDY312088
Thermoplastic Polyether Polyurethane Tags for M610 and M611
Etiketit, tarrat
These B-7643 cable tags contain no added halogens and offer excellent fire safety characteristics combined with minimal smoke emission.
  • Thermoplastic Polyether Polyurethane tags
  • Excellent chemical, oil, and water resistance
  • Operating temperature of −40 to +90 °C
  • Matte finish
Applications include data and telecommunications labelling, and wire and cable labelling.
Sertifikaatit: Halogen-free per IEC 61249-2-21 (2003-11), Adherence: SAE-AS81531-1998, Flame retardant: MIL-STD-202G.
Pakkaus: 50 tags per roll.