Label printers with Brady workstation lab suite, BBP® 12
Toimittaja: Brady
306866 306867
BRDY306866EA 964 EUR
BRDY306866 BRDY306867
Label printers with Brady workstation lab suite, BBP® 12
Tulostimet ja tulostimien lisävarusteet Label Printers
The Brady BBP® 12 label printers are compact, reliable, affordable entry level thermal transfer benchtop printer.
  • 300 dpi
  • Mono colour
  • Applications include asset identification, barcode printing, cable and wire identification, and laboratory identification
Ideal for customers requiring low volume printing on lots of different materials within the electrical, voice and datacom, LAB and specialized electronics markets. It allows you to print labels, tapes cable markers, sleeves and much more. The printer comes with the Brady Workstation Product and WireMore Product Information