Safety lockout hasp, blank
Toimittaja: Brady
225196 225198 265399
BRDY225196EA 107 EUR
BRDY225196 BRDY225198 BRDY265399
Safety lockout hasp, blank
Turvalukot
These blank lockout hasps can easily be customised with labels.
  • Made from aluminium
  • Available in several colours
  • Six padlocks can be applied
Max. shackle diameter: 8,00 mm
Min. shackle clearance: 20,00 mm