Codesoft Lite Software
Toimittaja: Brady
196181 196184 362918 362916
BRDY196181EA 566 EUR
BRDY196181 BRDY196184 BRDY362918 BRDY362916
Codesoft Lite Software
Ohjelmisto Laboratorio-ohjelmisto
Codesoft Lite label design software can be used for asset tracking, barcode labelling, and inventory and inspection labelling applications.
  • Only available as a single user license
  • Limited to 1 thermal printer
Languages availble include Arabic (Egypt), Czech, Danish, Dutch, English, Finnish, French, German, Hebrew, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Spanish, Swedish, and Turkish.
Compatible with BBP11, BBP12, BBP72, PR Plus, IP Series, PAM3000, PAM3600, THT 76mm Core, and TLS-PC LINK printers.