Compact cable safety padlocks
Toimittaja: Brady
195948 195949 195950 195951 195952 195953 146120 146121 146122 146123 146124 146125 146126 146127 146128 146129 146130 146131 146132 146133 146134 146135 195934 195935 195936 195937 195938 195939 195940 195941 195942 195943 195944 195945 195946 195947 195954 195955 195956 195957 195972 195973 195974 195975 195976 195977 195978 195979 195980 195981 195982 195983 195984 195985 195986 195987 195988 195989 195990 195991
BRDY195948EA 195 EUR
BRDY195948 BRDY195949 BRDY195950 BRDY195951 BRDY195952 BRDY195953 BRDY146120 BRDY146121 BRDY146122 BRDY146123 BRDY146124 BRDY146125 BRDY146126 BRDY146127 BRDY146128 BRDY146129 BRDY146130 BRDY146131 BRDY146132 BRDY146133 BRDY146134 BRDY146135 BRDY195934 BRDY195935 BRDY195936 BRDY195937 BRDY195938 BRDY195939 BRDY195940 BRDY195941 BRDY195942 BRDY195943 BRDY195944 BRDY195945 BRDY195946 BRDY195947 BRDY195954 BRDY195955 BRDY195956 BRDY195957 BRDY195972 BRDY195973 BRDY195974 BRDY195975 BRDY195976 BRDY195977 BRDY195978 BRDY195979 BRDY195980 BRDY195981 BRDY195982 BRDY195983 BRDY195984 BRDY195985 BRDY195986 BRDY195987 BRDY195988 BRDY195989 BRDY195990 BRDY195991
Compact cable safety padlocks
Turvalukot
Featuring an elongated, flexible cable, these padlocks are more versatile than standard safety padlocks, providing a multi-purpose solution. Suitable for energy isolation programs.
  • Traditional shackle is replaced by a 4,74 mm PVC encased steel cable
  • Padlock is non conductive to allow easy and safe usage in an electrical environment
  • Unique cylinder is insulated to protect workers from shocks when key is inserted
  • 5-pin precision-machined for more unique key cuts and better tamper resistance
  • Reserved, paracentric keyway provides optimal security; special