Handheld Wireless Palm Barcode Scanners: Bluetooth, 1D, 2D, QR Code, CR2700
Toimittaja: Brady
176686 176517 176518 176685
BRDY176686EA 1040 EUR
BRDY176686 BRDY176517 BRDY176518 BRDY176685
Handheld Wireless Palm Barcode Scanners: Bluetooth, 1D, 2D, QR Code, CR2700
Viivakoodijärjestelmät
Read wide and small barcodes easier with this ultra-fast barcode reader. Get unbeatable performance from this aggressive, omnidirectional scanner that reads even damaged barcodes.
  • Uses B27 batteries with built-in power gauge
  • IP65 seal rating against dust and liquid ingress
  • CodeShield® housing for disinfectant and chemical wipedown
  • Unequaled performance reading on curved and shiny surfaces
  • USB, Bluetooth®5 low energy (for enhanced data security, extended battery life, and less downtime)
Customisable visual, audible and haptic feedback with multipleMore Product Information
Order Now
TEKNISET TIEDOT
Environmentally Preferable
L×S×K 53,3×131,0×28,0 mm 53,3×132,1×27,9 mm