Self-Laminating Cryogenic Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Toimittaja: Brady
174342 311983 174347 173902 311985 174350 174371 174343 173702 173487 173905 173901 174351
BRDY174342EA 174 EUR
BRDY174342 BRDY311983 BRDY174347 BRDY173902 BRDY311985 BRDY174350 BRDY174371 BRDY174343 BRDY173702 BRDY173487 BRDY173905 BRDY173901 BRDY174351
Self-Laminating Cryogenic Polyester Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Etiketit, tarrat
Self-laminating polyester (B-461) is a clear film with a matte white printable zone ideal for cables, laboratory ID, or general labelling where graphics need to be protected.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance, and high/low temperature performance
  • Service temperature of −196 to +110 °C (−320 to +230 °F)
  • Matte finish
Ideal for laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printerMore Product Information