Retro Reflective All-Weather Labels for M610, M611 and BMP61
Toimittaja: Brady
174117 174130 174128 174129 174118
BRDY174117EA 229 EUR
BRDY174117 BRDY174130 BRDY174128 BRDY174129 BRDY174118
Retro Reflective All-Weather Labels for M610, M611 and BMP61
Etiketit, tarrat
Highly reflective B-584 material shines brightly when struck by light, which is ideal for applications where high visibility is critical.
  • Retro reflective sheeting and pressure sensitive acrylic adhesive meet Federal Spec L-S-300 for Reflectivity
  • Average outdoor durability is 4 to 6 years
  • Service temperature of −40 to +70 °C (−40 to +158 °F)
  • Rflective gloss finish
Labels are ideal for component and equipment labelling, facility identification, general identification, and safety identification applications. Perfect for labelling job site tools, hardMore Product Information