All-Weather Vinyl Labels for M610, M611, BMP61 Printers
Toimittaja: Brady
174158 174119 174136 174123 174140 174126 174159 174141 174122 174137 174163 174138 174121 174127 174157 174160 174120 174132 174156 174139 174162 174161 174124 174134 174135 174131 174133 174125 174142
BRDY174158EA 88.5 EUR
BRDY174158 BRDY174119 BRDY174136 BRDY174123 BRDY174140 BRDY174126 BRDY174159 BRDY174141 BRDY174122 BRDY174137 BRDY174163 BRDY174138 BRDY174121 BRDY174127 BRDY174157 BRDY174160 BRDY174120 BRDY174132 BRDY174156 BRDY174139 BRDY174162 BRDY174161 BRDY174124 BRDY174134 BRDY174135 BRDY174131 BRDY174133 BRDY174125 BRDY174142
All-Weather Vinyl Labels for M610, M611, BMP61 Printers
Etiketit, tarrat
This durable, low-shrink vinyl with ultra-aggressive adhesive is the perfect solution for identification on rough surfaces and in harsh environment areas.
  • Permanent adhesive sticks to surfaces that are smooth, rough, or powder coated
  • Will also stick to irregular surfaces or surfaces with low surface energy
  • Clean gloss finish is resistant to chemicals, grease, oil, and cleaning agents, ensuring identification stays legible
  • This material has an average outdoor durability of 8 to 10 years
  • Gloss finish
Vinyl tape is ideal for use in pipe marking,More Product Information