Solvent Resistant Polyester Labels for M710 and BMP71
Toimittaja: Brady
173308 173301 173309
BRDY173308EA 140 EUR
BRDY173308 BRDY173301 BRDY173309
Solvent Resistant Polyester Labels for M710 and BMP71
Etiketit, tarrat
High performance polyester (B-488) withstands numerous solvents and variable temperatures when applied to various surfaces, including stainless steel and polypropylene.
  • Matte white polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • Ideal for general and bar code labelling on flat, smooth surfaces
  • Service temperature of −40 to +160 °C (−40 to +320 °F)
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspectionMore Product Information