FreezerBondz™ Ultra-Thin Polyester Laboratory Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Toimittaja: Brady
170781 170897 170898 170892 170901 170777 170893 170895 170896 170900
BRDY170781EA 130 EUR
BRDY170781 BRDY170897 BRDY170898 BRDY170892 BRDY170901 BRDY170777 BRDY170893 BRDY170895 BRDY170896 BRDY170900
FreezerBondz™ Ultra-Thin Polyester Laboratory Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketit, tarrat
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady handheld label printer.
  • Ideal for laboratory use on curved surfaces including test tubes, vials, and straw pipettes
  • Withstands chemicals, abrasions, and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
  • Aggressive, permanent adhesive
  • Matte finish
Matte finish,More Product Information