Indoor Outdoor Vinyl Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Toimittaja: Brady
170814 170813 170815 170816 170817 170818 170819 170820 170821 170823 170824 170825 170826 170827 170828 170829 170830 170831 170832 170833 170834 170835 170836 170837 170838 170839 170840 170841 170842 170844 170845 170939 170940 170941 170942 170943 170945 170946 170947 170948 170950
BRDY170814EA 47.5 EUR
BRDY170814 BRDY170813 BRDY170815 BRDY170816 BRDY170817 BRDY170818 BRDY170819 BRDY170820 BRDY170821 BRDY170823 BRDY170824 BRDY170825 BRDY170826 BRDY170827 BRDY170828 BRDY170829 BRDY170830 BRDY170831 BRDY170832 BRDY170833 BRDY170834 BRDY170835 BRDY170836 BRDY170837 BRDY170838 BRDY170839 BRDY170840 BRDY170841 BRDY170842 BRDY170844 BRDY170845 BRDY170939 BRDY170940 BRDY170941 BRDY170942 BRDY170943 BRDY170945 BRDY170946 BRDY170947 BRDY170948 BRDY170950
Indoor Outdoor Vinyl Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketit, tarrat
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label printer.
  • Outdoor durable
  • Low temperature resistant
  • Fuel and oil resistant
  • High adhesion
  • Abrasion resistant
  • Gloss finish
Ideal for labelling pipes, equipment, shelves and other industrial surfaces. Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime. Made with high-tack, permanent acrylic adhesive for use onMore Product Information