Label printers EMEA, M211
Toimittaja: Brady
170381 170390
BRDY170381EA 166 EUR
BRDY170381 BRDY170390
Label printers EMEA, M211
Tulostimet ja tulostimien lisävarusteet Label Printers
Seamlessly design, preview and print labels from your familiar smartphone and swiftly connect to a reliable, industrial grade printer. No other phone-driven label printer today matches the label design possibilities of the M211 Label Printer.
  • Use the M211 Label Printer, Express Labels Mobile App and a phone to create and print labels
  • Tough, industrial labeler designed to survive and thrive on any worksite.
  • Get more done all day long with 300 label prints from one full battery charge
  • Fast and dependable Bluetooth connection
  • Design, preview and print even