Labels for BBP33 Printers, B-8591
Toimittaja: Brady
152941 152931 152946 152936 152937 152932 152942 152947
BRDY152941EA 455 EUR
BRDY152941 BRDY152931 BRDY152946 BRDY152936 BRDY152937 BRDY152932 BRDY152942 BRDY152947
Labels for BBP33 Printers, B-8591
Etiketit, tarrat
B-8591 outdoor, harsh-environment labels resist UV, abrasions, high temperatures, fluids and washdowns, and are an ideal print-on-demand thermal transfer label solution.
  • No overlaminate means durability is drastically improved without adding an overlaminate over the print
  • Abrasion-, chemical-, corrosion-, fade-, humidity-, UV-, weather-resistant
  • Quick and easy application with pressure-sensitive adhesive
  • Meets regulatory specifications for halogen-free applications
  • A ribbon is required to print this material
12+ year outdoor durability reducesMore Product Information