Labelling tapes, thermal transfer printable tapes
Toimittaja: Brady
150608 150609 150610 150611 150612 150613 150614 150615
BRDY150608EA 349 EUR
BRDY150608 BRDY150609 BRDY150610 BRDY150611 BRDY150612 BRDY150613 BRDY150614 BRDY150615
Labelling tapes, thermal transfer printable tapes
Etiketit, tarrat
Continuous tapes for printers.
  • Choice of repositionable rubber or permanent acrylic adhesives
  • Continuous tape is not cut to length, it gets cut by the printer
  • This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves
Vinyl labelling tapes feature a repositionable, rubber based adhesive and semi-gloss finish. Vinyl tapes are ideal for general identification, laboratory labelling, and wire and cable labelling applications. These tapes should be used with IP-R6206 and R6206 ribbons, and areMore Product Information