Labels for BradyJet J2000 Lab printer
Toimittaja: Brady
150005 150006 150007 150011 150015 150016 150018 150019 150020 150022 150023 150024 150025 150028 150033 150163 150542 150545 150543 150541 150544 150547
BRDY150005EA 201 EUR
BRDY150005 BRDY150006 BRDY150007 BRDY150011 BRDY150015 BRDY150016 BRDY150018 BRDY150019 BRDY150020 BRDY150022 BRDY150023 BRDY150024 BRDY150025 BRDY150028 BRDY150033 BRDY150163 BRDY150542 BRDY150545 BRDY150543 BRDY150541 BRDY150544 BRDY150547
Labels for BradyJet J2000 Lab printer
Etiketit, tarrat
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
  • Designed to stay attached and remain legible
  • Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.