SafeKey lockout padlocks, nylon
Toimittaja: Brady
150294 150234 150250 150251 150275 150320 150321 150343 150367 150368 150270 150225 150337 150316 150224 150336 150362 150269 150292 150228 150229 150246 150271 150366 150311 150265 150334 150221 150351 150310 150350 150309 150308 150222 150223 150267 150268 150317 150340 150341 150364 150233 150247 150248 150249 150274 150295 150296 150319 150357 150230 150231 150232 150272 150273 150318 150342 150365 150314 150315 150353 150291 150290
BRDY150294EA 84.8 EUR
BRDY150294 BRDY150234 BRDY150250 BRDY150251 BRDY150275 BRDY150320 BRDY150321 BRDY150343 BRDY150367 BRDY150368 BRDY150270 BRDY150225 BRDY150337 BRDY150316 BRDY150224 BRDY150336 BRDY150362 BRDY150269 BRDY150292 BRDY150228 BRDY150229 BRDY150246 BRDY150271 238-0179 BRDY150311 BRDY150265 BRDY150334 BRDY150221 BRDY150351 BRDY150310 BRDY150350 BRDY150309 BRDY150308 BRDY150222 BRDY150223 BRDY150267 BRDY150268 BRDY150317 BRDY150340 BRDY150341 BRDY150364 BRDY150233 BRDY150247 BRDY150248 BRDY150249 BRDY150274 BRDY150295 BRDY150296 BRDY150319 BRDY150357 238-0182 238-0178 238-0186 238-0183 238-0181 238-0180 238-0184 238-0185 BRDY150314 BRDY150315 BRDY150353 BRDY150291 BRDY150290
SafeKey lockout padlocks, nylon
Turvalukot
These SafeKey nylon padlocks are the ultimate locks for all types of safety and security needs.
  • Lightweight with a ribbed design for easy gripping
  • Ideal for electrical applications and enclosures
  • Key releases only upon lock closure
  • Preferred for most indoor lockout applications
  • Engraving options on front, back, left side, right side, and/or key
  • Key retaining (Key won’t release until the padlock shackle is securely closed)
  • Non-conductive
The new design and innovative keying mechanism provides more unique key possibilities than a conventional standardMore Product Information