SafeKey compact lockout padlocks, nylon
Toimittaja: Brady
150183 150184 150185 150186 150187 150188 150189 150190 150191 150192 150193 150194 150195 150196 150197 150207 150208 150209 150210 150211 150212 150213 150214 150215 150180 150181 150182
BRDY150183EA 22.2 EUR
BRDY150183 BRDY150184 BRDY150185 BRDY150186 BRDY150187 BRDY150188 BRDY150189 BRDY150190 BRDY150191 BRDY150192 BRDY150193 BRDY150194 BRDY150195 BRDY150196 BRDY150197 BRDY150207 BRDY150208 BRDY150209 BRDY150210 BRDY150211 BRDY150212 BRDY150213 BRDY150214 BRDY150215 BRDY150180 BRDY150181 BRDY150182
SafeKey compact lockout padlocks, nylon
Turvalukot
These SafeKey nylon compact padlocks are a small and lightweight range of padlocks suitable for use in energy isolation programs or in small spaces.
  • Lightweight with a ribbed design for easy gripping
  • Ideal for electrical applications and enclosures
  • Key releases only upon lock closure
  • Preferred for most indoor lockout applications
  • Engraving options on front, back, left side, right side, and/or key
  • Non-conductive
The new design and innovative keying mechanism provides more unique key possibilities than a conventional standard safety padlock, withoutMore Product Information