Indoor/outdoor labelling tapes for BBP3x printers
Toimittaja: Brady
142027 142029 142030 142032 142034 142035 142009 141993 141991 142007 142008 141992 142024 142025 141997 142013 142017 142021 142023 142001 142002 142031 142028 142005 142003 142026 142004 142006 142010 142011 142012 142014 142015 142016 142018 142019 142022 142033 142036 142000 142020 141987 141988 141989 141990 141994 141995 141996 141998 141999 146042 146043
817-0344EA 567 EUR
817-0344 BRDY142029 BRDY142030 817-0345 BRDY142034 BRDY142035 BRDY142009 BRDY141993 817-0334 817-0338 BRDY142008 BRDY141992 BRDY142024 BRDY142025 BRDY141997 BRDY142013 BRDY142017 BRDY142021 817-0343 BRDY142001 BRDY142002 BRDY142031 BRDY142028 BRDY142005 817-0337 BRDY142026 BRDY142004 BRDY142006 BRDY142010 817-0339 BRDY142012 BRDY142014 817-0341 BRDY142016 BRDY142018 817-0342 BRDY142022 BRDY142033 BRDY142036 BRDY142000 BRDY142020 817-0333 BRDY141988 BRDY141989 BRDY141990 BRDY141994 817-0335 BRDY141996 BRDY141998 817-0336 BRDY146042 BRDY146043
Indoor/outdoor labelling tapes for BBP3x printers
Etiketit, tarrat
Durable, low-shrink vinyl with our most aggressive adhesive. Conforms to irregular, curved, rough and highly textured surfaces (painted cinder blocks, uneven wood, textured plastics, paper-jacketed pipes, powder-coated surfaces). Adheres to 'difficult' low-surface energy items (PVC piping, blow-molded equipment cases, ABS plastics, recycled plastics).
Length: 30,0 m
Different colours available.