Thermal transfer printer labels, type B-499
Toimittaja: Brady
217075 217073 217082 062378 217081 062358 030542 019260 062375 062377 062376 217068 030543 116376 116379 217084 217066 116382 116368 116370 116384 217067 804776 217069
BRDY217075EA 455 EUR
BRDY217075 BRDY217073 BRDY217082 BRDY062378 BRDY217081 BRDY062358 BRDY030542 BRDY019260 BRDY062375 BRDY062377 BRDY062376 BRDY217068 BRDY030543 BRDY116376 BRDY116379 BRDY217084 BRDY217066 BRDY116382 BRDY116368 BRDY116370 BRDY116384 BRDY217067 BRDY804776 BRDY217069
Thermal transfer printer labels, type B-499
Etiketit, tarrat
Printable, nylon cloth labels for thermal transfer printers.
  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves
This material is UL recognised with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.