Group lockout boxes for wall-mounting
Toimittaja: Brady
105714 105715
BRDY105714EA 141 EUR
BRDY105714 BRDY105715
Group lockout boxes for wall-mounting
Turvalukot
These lockout boxes are made of durable, powder-coated steel with key slot on top.
  • Clear Lexan window slides out when locks are removed
  • Comes with 7 or 15 key pegs that hold 5 keys each and ID labels to simplify key management
  • Pre-drilled holes and hardware included for quick mounting
Legend language: English