Group lockout hasps, labelled
Toimittaja: Brady
065960 065970 065961 065962 065963 065964 065967 065971 065972 065973 065974 065975
BRDY065960EA 162 EUR
BRDY065960 BRDY065970 BRDY065961 BRDY065962 BRDY065963 BRDY065964 BRDY065967 BRDY065971 BRDY065972 BRDY065973 BRDY065974 BRDY065975
Group lockout hasps, labelled
Turvalukot
These labelled group lockout hasps are constructed from tough 5052 anodised aluminium alloy.
  • 6 padlocks can be applied
  • Include tamperproof locking tabs
  • Write-on label surface accept pencils, pen or marker
  • Pen and pencil markings can be easily erased, allowing repeated use
  • Lock hole diameter: 8 mm
Toimitustiedot: The assorted pack contains one each of the following colours: red, silver, blue, green and gold.