Labels for BBP85 and PowerMark printers, type B-595
Toimittaja: Brady
013562 013563 013610 013611 013612 013549 013601 013603 013605
BRDY013562EA 374 EUR
BRDY013562 BRDY013563 BRDY013610 BRDY013611 BRDY013612 BRDY013549 BRDY013601 BRDY013603 BRDY013605
Labels for BBP85 and PowerMark printers, type B-595
Etiketit, tarrat
These labeling tapes are made of vinyl film with permanent acrylic adhesive.
  • Sticks to highly textured surfaces
  • Adheres to difficult low surface energy items
  • Gloss finish
B-595 vinyl tapes will adhere to difficult low surface energy items, including PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces, and equipment made from recycled plastics. The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes, and paper-jacketed pipes.