Kanyyli, IV-kanyyli, Vasofix® Safety
Toimittaja: B. Braun
4268130S-01 4268156S-01 4268091S-01 4269071S-01 4268334S-01 4269136S-01 4269110S-01
BRAU4268130S-01EA 109 EUR
BRAU4268130S-01 BRAU4268156S-01 BRAU4268091S-01 BRAU4269071S-01 BRAU4268334S-01 BRAU4269136S-01 BRAU4269110S-01
Kanyyli, IV-kanyyli, Vasofix® Safety
Katetrit
IV catheters with injection port and self-activating needlestick protection. Transparent segments provide confirmation of successful catheter placement when withdrawing the needle.
  • Easy puncture with minimum effort of catheter insertion
  • Self-activating protection against needlestick injuries and related infection
  • Needle tip automatically covered after use
  • Simplicity – Looks and feels like a standard cannula
  • Sharp backcut bevel for minimal puncture trauma