Cryopreservation freeze media, CryoStor®
Toimittaja: Charles River Laboratories Cell Solutions
BPM202102 BPM205102 BPM210373 BPM210102 BPM210202
202102.EA 0 EUR
202102. BPM205102 210373. 210102. 210202.
Cryopreservation freeze media, CryoStor®
Solujen syväjäädytysliuokset
CryoStor® cryopreservation freeze media products have been designed to mitigate temperature-induced molecular cell stress responses during freezing and thawing.
CryoStor® freeze media products are intended for cryopreservation of biologics at −80 to −196 °C and are based on the novel HypoThermosol® formula.
All CryoStor® products are pre-formulated with USP grade DMSO, a permeant solute cryoprotective agent which helps mitigate damage from the formation of intracellular ice.
Across a broad spectrum of cell types, CryoStor® products have proven much more effective in reducing post-preservation necrosis and apoptosis as compared to commercial and home-brew isotonic and extracellular formulations. This enables greatly improved post-thaw cell yield, viability, and recovery.
Sertifikaatit: Meet USP Sterility and USP Endotoxin testing standards, and are manufactured under cGMP.
Varoitus: Research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.