Anti-C15ORF52 Rabbit Polyclonal Antibody (Cy7®)

Toimittaja: Bioss
BS-9620R-CY7
BOSSBS-9620R-CY7EA 1350 EUR
BOSSBS-9620R-CY7
Anti-C15ORF52 Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
C15orf52

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: C15ORF52
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR