Anti-LRP15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Toimittaja: Bioss
BS-9509R-A680
BOSSBS-9509R-A680EA 893 EUR
BOSSBS-9509R-A680
Anti-LRP15 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Vasta-aineet
Leucine-rich repeats (LRRs) are 20-29 amino acid motifs that mediate protein-protein interactions. The primary function of these motifs is to provide a versatile structural framework for the formation of these protein-protein interactions. LRRs are present in a variety of proteins with diverse structure and function, including innate immunity and nervous system development. Several human diseases are associated with mutation in the genes encoding LRR-containing proteins. The leucine-rich repeat-containing protein 3B (LRRC3B), also designated LRP15, is a 259 amino acid protein that contains 3More Product Information