Anti-PAPOL A+B+G Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-9435R-A750
BOSSBS-9435R-A750EA 893 EUR
BOSSBS-9435R-A750
Anti-PAPOL A+B+G Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
Polyadenylation of the 3-prime ends of eukaryotic mRNAs is a key event that takes place in the nucleus during maturation of mRNA. The reaction includes endoribonucleolytic cleavage of the pre-RNA at the poly(A) site that leads to synthesis of the poly(A) tail at the 3-prime end of the upstream cleavage product. The poly(A) polymerase (PAP) is required The adenosine addition reaction depends on poly(A) polymerase (PAP) activity. The testis express PAP-beta (TPAP) in the cytoplasm of spermatogenic cells. The adenosine addition function of PAP-beta plays a critical role in male germ cellMore Product Information