Anti-TRPM4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Toimittaja: Bioss
BS-9051R-A555
BOSSBS-9051R-A555EA 893 EUR
BOSSBS-9051R-A555
Anti-TRPM4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Vasta-aineet
Calcium-activated non selective (CAN) cation channel that mediates membrane depolarization. While it is activated by increase in intracellular Ca(2+), it is impermeable to it. Mediates transport of monovalent cations (Na(+) >K(+) >Cs(+) >Li(+)), leading to depolarize the membrane. It thereby plays a central role in cadiomyocytes, neurons from entorhinal cortex, dorsal root and vomeronasal neurons, endocrine pancreas cells, kidney epithelial cells, cochlea hair cells etc. Participates in T-cell activation by modulating Ca(2+) oscillations after T lymphocyte activation, which isMore Product Information