Anti-GABR B3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Toimittaja: Bioss
BS-8544R-A680
BOSSBS-8544R-A680EA 877 EUR
BOSSBS-8544R-A680
Anti-GABR B3 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Vasta-aineet
Component of the heteropentameric receptor for GABA, the major inhibitory neurotransmitter in the vertebrate brain. Functions also as histamine receptor and mediates cellular responses to histamine. Functions as receptor for diasepines and various anesthetics, such as pentobarbital; these are bound at a separate allosteric effector binding site. Functions as ligand-gated chloride channel.

Type: Primary
Antigen: GABR B3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-130/473
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR