Anti-BTBD10 Rabbit Polyclonal Antibody (Cy7®)
Toimittaja: Bioss
BS-8398R-CY7
BOSSBS-8398R-CY7EA 900 EUR
BOSSBS-8398R-CY7
Anti-BTBD10 Rabbit Polyclonal Antibody (Cy7®)
Vasta-aineet
The BTB (broad-complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. BTBD10 (BTB (POZ) domain containing 10), also known as GMRP1, is a ubiquitously expressed nuclear protein found at highest levels in adult testis, brain and small intestine and weakly expressed in colon, lung, liver, kidney,More Product Information