Anti-BTBD10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Toimittaja: Bioss
BS-8398R-A750
BOSSBS-8398R-A750EA 893 EUR
BOSSBS-8398R-A750
Anti-BTBD10 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Vasta-aineet
The BTB (broad-complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and zinc finger) domain, is an N-terminal homodimerisation domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. BTBD10 (BTB (POZ) domain containing 10), also known as GMRP1, is a ubiquitously expressed nuclear protein found at highest levels in adult testis, brain and small intestine and weakly expressed in colon, lung, liver, kidney,More Product Information