Anti-LRRC41 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Toimittaja: Bioss
BS-8362R-A488
BOSSBS-8362R-A488EA 893 EUR
BOSSBS-8362R-A488
Anti-LRRC41 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Vasta-aineet
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli LRRC41
Antigeenin nimi Leucine Rich Repeat Containing 41
Environmentally Preferable
Antigeenin synonyymit PP7759|MUF1
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Alexa Fluor® 488
Reaktiivisuus Human,Rat,Mouse
Isäntä Rabbit
Geeni ID 10489
Isotyyppi IgG
ImmunoKemia Yes
Lomake liquid
Puhdistus Purified by Protein A
Pitoisuus 1 μg/μl
Ristiadsorptio No
Säilytyspuskuri Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Säilytyslämpötila Store at –20 °C for 12 months
Kuljetuslämpötila 4°C