Anti-LRRC41 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Toimittaja: Bioss
BS-8362R-A350
BOSSBS-8362R-A350EA 893 EUR
BOSSBS-8362R-A350
Anti-LRRC41 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Vasta-aineet
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information