Anti-DPYSL2 Rabbit Polyclonal Antibody (Cy5.5®)
Toimittaja: Bioss
BS-8284R-CY5.5
BOSSBS-8284R-CY5.5EA 930 EUR
BOSSBS-8284R-CY5.5
Anti-DPYSL2 Rabbit Polyclonal Antibody (Cy5.5®)
Vasta-aineet
Collapsin response mediator proteins (CRMPs) are cytosolic phosphoproteins involved in neuronal differentiation and axonal guidance. CRMP2 was previously shown to mediate the repulsive effect of Sema3A on axons and to participate in axonal specification. The CRMPs appear to play a complex role in axon growth as well as microtubule dynamics and axon induction. CRMPs localize to the lamellipodia and filopodia of axonal growth cones, suggesting a role in axon guidance. Moreover, CRMP2 is upregulated after axotomy, and appears to increase the formation of axon-type processes from hippocampalMore Product Information